missing translation for 'onlineSavingsMsg'
Learn More

Zinc Citrate Tribasic Dihydrate, purum, ≥98% (KT), Honeywell Fluka™

Product Code. 15687580 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15687580 100 g Poly Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15687580 Supplier Honeywell-Fluka Supplier No. 96494100G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum,≥98.0% (calc. based on dry substance, KT)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 334°C
CAS 5990-32-9
Color White
Quantity 100 g
Molecular Formula C12H14O16Zn3
Linear Formula (C6H5O7)2Zn3 · 2H2O
MDL Number MFCD00150587
UN Number NONH for all modes of transport
Beilstein 3819614
Synonym zinc citrate dihydrate, unii-021ibv57il, acmc-20aj8r, citric acid zinc salt dihydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, zinc salt 2:3 , dihydrate, trizinc 2+ dicitrate dihydrate, zinc citrate, dihydrate, reagent, trizinc, ion zn2+ dicitrate dihydrate, zinc 2-hydroxypropane-1,2,3-tricarboxylate-water 3/2/2, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, zinc salt, hydrate 2:3:2
InChI Key OXAGUGIXGVHDGD-UHFFFAOYSA-H
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Zn+2].[Zn+2].[Zn+2]
IUPAC Name trizinc;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
Molecular Weight (g/mol) 610.368
PubChem CID 16213234
Formula Weight 610.4g/mol
Percent Purity ≥98% (calc. based on dry substance, KT)
Packaging Poly Bottle
Appearance Powder
Chemical Name or Material Zinc citrate tribasic dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.