missing translation for 'onlineSavingsMsg'
Learn More

Zinc copper couple

Product Code. 10591301 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10591301 25 g Glass bottle 25g
10369100 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10591301 Supplier Thermo Scientific Acros Supplier No. 271440250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 53801-63-1 | CuZn | 128.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53801-63-1
Molecular Formula CuZn
Molecular Weight (g/mol) 128.93
MDL Number MFCD00084851
InChI Key TVZPLCNGKSPOJA-UHFFFAOYSA-N
Synonym zinc-copper couple, cu.zn, zinc copper, copper, compound with zinc 1:1, zinc-copper, brass alloy, zn copper, copper-zinc alloy, zinc copper couple, cu-zn
PubChem CID 10290809
IUPAC Name copper;zinc
SMILES [Cu].[Zn]

Specifications

Color Gray to Black
Quantity 25 g
Packaging Glass bottle
Form Powder
Linear Formula ZnCu
Fieser 14,309; 15,367; 16,331; 17,407
Chemical Name or Material Zinc copper couple
compliance-icons compliance-icons
Product Identifier
  • Zinc copper couple
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.