missing translation for 'onlineSavingsMsg'
Learn More

Zinc gluconate hydrate, 97%

Product Code. 11406206
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406206 100 g 100g
11416206 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11406206 Supplier Thermo Scientific Alfa Aesar Supplier No. B23022.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4468-02-4 | C4H7NO | 85.11 g/mol

Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc gluconate hydrate is used as a food additive, a pharmaceutical intermediate.

Solubility
Soluble in water 100g/ l.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4468-02-4
MDL Number MFCD00868110

Specifications

pH 6.5
Odor Odorless
Molecular Formula C12H22O14Zn
Quantity 100 g
Merck Index 14,4456
Synonym zinc ii gluconate hydrate, zinc 2+ bis d-gluconate hydrate
Solubility Information Soluble in water 100g/ l.
InChI Key WHMDKBIGKVEYHS-IYEMJOQQSA-L
SMILES [Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
IUPAC Name zinc;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate;hydrate
Molecular Weight (g/mol) 455.67
PubChem CID 131674739
Formula Weight 455.69 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Zinc gluconate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.