missing translation for 'onlineSavingsMsg'
Learn More

Zinc(II) acetylacetonate, ca. 25% Zn

Product Code. 10569833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10569833 100 g Glass bottle 100g
10176863 5 g Glass bottle 5g
10783245 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10569833 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 388771000
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14024-63-6 | C10H14O4Zn | 263.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-63-6
Molecular Formula C10H14O4Zn
Molecular Weight (g/mol) 263.60
MDL Number MFCD00000035
InChI Key CYDXJXDAFPJUQE-FDGPNNRMSA-L
Synonym zinc acetylacetonate hydrate, zinc 2,4-pentanedionate, hydrate, zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
PubChem CID 131675103
SMILES [Zn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Boiling Point 129.0°C to 131.0°C (10.0mmHg)
Melting Point 124.0°C to 126.0°C
Color White to Ivory
Physical Form Powder
Quantity 100 g
Solubility Information Solubility in water: 6.9g/L (20°C)
IUPAC Name zinc(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Formula Weight 263.59
Percent Purity ∼25% (Zn)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Zinc(II) acetylacetonate
Show More Show Less
compliance-icons
Product Identifier
  • Zinc(II) acetylacetonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.