missing translation for 'onlineSavingsMsg'
Learn More

Zinc iodide, 98+%, pure

Product Code. 10686782 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 10686782

Brand: Thermo Scientific Acros 208062500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10139-47-6 | I2Zn | 319.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10139-47-6
Molecular Formula I2Zn
Molecular Weight (g/mol) 319.19
MDL Number MFCD00011299

Specifications

Melting Point 446°C
Color Beige-Yellow to White
Physical Form Hygroscopic Crystalline Powder or Crystals
Quantity 250 g
Assay Percent Range 98% min. (Argentometry)
Linear Formula ZnI2
Fieser 01,1294; 05,765; 10,462; 11,604; 12,576; 13,88
Merck Index 15, 10339
Solubility Information Solubility in water: 4500g/L (20°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
Formula Weight 319.19
Percent Purity ≥98%
Grade Pure
Packaging Glass bottle
Specific Gravity 4.74
Density 4.7400g/mL
Chemical Name or Material Zinc iodide, 98+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.