missing translation for 'onlineSavingsMsg'
Learn More

Zinc nitrate hexahydrate, 98%, extra pure

Product Code. 10286980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10286980 5 g Glass Bottle 5g
10036103 250 g Plastic bottle 250g
10276980 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10286980 Supplier Thermo Scientific Acros Supplier No. 211660050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10196-18-6 | H12N2O12Zn | 297.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10196-18-6
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 36.0°C
Color White
Physical Form Adhering Crystals
Quantity 5 g
Assay Percent Range 98%
Linear Formula Zn(NO3)2·6H2O
Merck Index 14,10144
Solubility Information Solubility in water: 1800g/L (20°C). Other solubilities: freely soluble in alcohol
IUPAC Name zinc;dinitrate;hexahydrate
Formula Weight 297.46
Percent Purity 98%
Grade Extra Pure
Packaging Glass Bottle
Chemical Name or Material Zinc nitrate hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.