missing translation for 'onlineSavingsMsg'
Learn More

Zinc nitrate hexahydrate, 99% (metals basis)

Product Code. 11331679
Change view
Click to view available options
Quantity:
500 g
2 kg
10 kg
Unit Size:
10kg
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331679 10 kg 10kg
11311679 500 g 500g
11321679 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331679 Supplier Thermo Scientific Alfa Aesar Supplier No. 012313.A9
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10196-18-6 | H12N2O12Zn | 297.48 g/mol

Zinc nitrate hexahydrate is used in the synthesis of coordination polymers. It is utilized in the preparation of zinc oxide based nanowires. It is also used as a mordant in dyeing. It is involved in the preparation of a new microporous, organically templated zinc phosphate, which finds application as catalysts and other applications involving microporosity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10196-18-6
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
IUPAC Name zinc(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point ∽36°C
Physical Form Crystalline
Quantity 10 kg
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in water,alcoholSoluble in water and ethanol.
Formula Weight 297.47 (189.38 Anhydrous)
Sensitivity Hygroscopic
Density 2.07
Chemical Name or Material Zinc nitrate hexahydrate
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Zinc nitrate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.