missing translation for 'onlineSavingsMsg'
Learn More

Zinc Nitrate Hexahydrate, TRC

Product Code. 30456882
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456882 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456882 Supplier Toronto Research Chemicals Supplier No. TRCZ4398001G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10196-18-6
Quantity 1 g
Molecular Formula H12N2O12Zn
Synonym Nitric Acid Zinc Salt Hydrate, Nitric Acid Zinc Salt Hexahydrate, Zinc Dinitrate Hexahydrate
InChI Formula InChI=1S/2NO3.6H2O.Zn/c2*2-1(3)4;;;;;;;/h;;6*1H2;/q2*-1;;;;;;;+2
SMILES O.O.O.O.O.O.[Zn+2].[O-][N+](=O)[O-].[O-][N+](=O)[O-]
IUPAC Name zinc;dinitrate;hexahydrate
Molecular Weight (g/mol) 297.51
Formula Weight 295.9682
Chemical Name or Material Zinc Nitrate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.