missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, 99.99% (metals basis)

Product Code. 11395039
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395039 250 g 250g
11385039 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395039 Supplier Thermo Scientific Alfa Aesar Supplier No. 087812.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1314-13-2 | OZn | 81.38 g/mol

Zinc oxide is used as a sun screener. It serves as an additive in products such as cigarette filters, plastics, anticorrosive coatings, ceramics, glass, cement, lubricants, ointments, adhesives, sealants, pigments, batteries, ferrites, fire retardants, and first-aid tapes. It plays an important role in textiles as a UV-absorber. As a wide-bandgap semiconductor with favorable properties, it has the potential for use in transparent electrodes, in nanorod sensors, in liquid crystal displays, as thin-film transistors and light emitting diodes, and in heat-protecting windows.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975°C
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Insoluble in water. Soluble in dilute acetic acid,mineral acids,ammonia,ammonium carbonate and fixed alkalihydroxide solution.
Formula Weight 81.37
Density 5.606
Chemical Name or Material Zinc oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.