missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, ACS reagent

Product Code. 10311261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
500g
5g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10311261 1 kg Plastic bottle 1kg
10720492 5 g Glass Bottle 5g
10390064 100 g Plastic bottle 100g
10701915 500 g Plastic bottle 500g
11914141 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10311261 Supplier Thermo Scientific Acros Supplier No. 422740010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1314-13-2 | OZn | 81.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975.0°C
Color White
Physical Form Powder
Quantity 1 kg
Linear Formula ZnO
Fieser 01,1294; 05,622
Merck Index 15,10347
Solubility Information Solubility in water: 1.6 mg/l (29°C). Other solubilities: soluble in acids and alkalies, soluble in diluted acetic or mineral acids, ammoni, ammonium carbonate, fixed alkali hydroxide solutio, insoluble in alcohol
Formula Weight 81.38
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 5.6
Density 5.6000g/mL
Chemical Name or Material Zinc oxide
Show More Show Less
compliance-icons
Product Identifier
  • Zinc oxide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Zinc oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.