missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Oxide, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Shop All Fisher Chemical ProductsSpecifications
Specifications
| Melting Point | 1975°C |
| Color | White |
| pH | 7 |
| Physical Form | Powder/Solid |
| Quantity | 500 g |
| Formula Weight | 81.38g/mol |
| Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Zinc oxyde
- Warning
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?