missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, heavy, 98%

Product Code. 11433267
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433267 1000 g 1000g
11443267 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433267 Supplier Thermo Scientific Alfa Aesar Supplier No. A16188.0B
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1314-13-2 | OZn | 81.38 g/mol

Mainly used in coating auxiliary agents, electronics chemicals, leather auxiliary agents, paper chemicals, petroleum additives, plastic auxiliary agents, rubber auxiliary agents,surfactants, textile auxiliary agents. Nanometer zinc oxide can also be used for high-grade ceramic material, can reduce the sintering temperature, achieving the purpose of saving energy. Used as the sulfuration active agents and reinforcing agents for various natural rubber and compound rubber products as well as the colorants and filling agents of white glue. It can also be used as additives to improve textile, textile ultraviolet resistant function and self-cleaning ability, can also be used in feed industry additive

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly used in coating auxiliary agents, electronics chemicals, leather auxiliary agents, paper chemicals, petroleum additives, plastic auxiliary agents, rubber auxiliary agents,surfactants, textile auxiliary agents. Nanometer zinc oxide can also be used for high-grade ceramic material, can reduce the sintering temperature, achieving the purpose of saving energy. Used as the sulfuration active agents and reinforcing agents for various natural rubber and compound rubber products as well as the colorants and filling agents of white glue. It can also be used as additives to improve textile, textile ultraviolet resistant function and self-cleaning ability, can also be used in feed industry additive

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong alkaline earth metal. Alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975°C
Quantity 1000 g
UN Number UN3077
Merck Index 14,10147
Solubility Information Insoluble in water.
Formula Weight 81.37
Percent Purity 98%
Odor Odorless
Density 5.606
Chemical Name or Material Zinc oxide, heavy

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.