missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide, NanoArc™ ZN-0605

Product Code. 15452467
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15452467 500 g 500g
15442467 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15452467 Supplier Thermo Scientific Alfa Aesar Supplier No. 045849.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1314-13-2 | OZn | 81.38 g/mol

Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis. Used in electronic industry, instrumental industry, manufacture of electrical device, image recorder, rheostat, phosphor. Used as a sun screening agent in cosmetics, antibacterial and health protection antiager; UV protection; Piezoelectrics; High-temperature lubricant in gas turbine engines; Flame retardant; Adsorption; Dental cements; Environmental remediation; Gas sensors; Photocatalytic decontamination; Attenuation of ultraviolet light; Demilitarization of chemical and biological warfare agents; Cosmetics and cosmeceuticals; Electrodes for solar cells; Varistors; Pigments for paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975°C
Color White
Physical Form Powder
Quantity 500 g
Linear Formula ZnO
UN Number UN3077
Merck Index 14,10147
Solubility Information Insoluble in water.
Formula Weight 81.37
Odor Odorless
Chemical Name or Material Zinc oxide NanoArc™ ZN-2605
Show More Show Less
compliance-icons
Product Identifier
  • Zinc oxide, NanoArc ZN-0605
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Zinc oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.