missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, puriss. p.a., ACS Reagent, ≥99% (KT), Honeywell Fluka™

Product Code. 15657580 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
250g
500g
This item is not returnable. View return policy

Product Code. 15657580

Brand: Honeywell-Fluka 96479500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (KT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point ∼1975°C
Color White
pH ∼7 (at 20°C)
Quantity 500 g
Linear Formula ZnO
UN Number UN3077
Formula Weight 81.39g/mol
Percent Purity ≥99% (KT)
Packaging Poly Bottle
Appearance Powder
Density 5.6g/cm3 at 20°C
Chemical Name or Material Zinc oxide
Show More Show Less
compliance-icons
Product Identifier
  • Zinc oxide
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : zinc oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.