missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.9% (metals basis)

Product Code. 15474845
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474845 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15474845 Supplier Thermo Scientific Alfa Aesar Supplier No. 041123.KS
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1314-13-2 | OZn | 81.38 g/mol

Zinc oxide is used as an additive in numerous materials and products including rubbers, plastics, ceramics, glass, cement, lubricants, paints, ointments, adhesives, sealants, pigments, foods, batteries, ferrites, fire retardants, and first-aid tapes. It is also used in oil, greases, ferrites, cosmetics, pharmaceuticals, medical, UV coatings, lacquers, ceramics and textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White to Yellow
Quantity 1 Ea.
CAS 1314-13-2
Odor Odorless
Molecular Formula OZn
Linear Formula ZnO
MDL Number MFCD00011300
UN Number UN3077
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
Solubility Information Soluble in diluted acid. Insoluble in water and alcohol.
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILES O=[Zn]
IUPAC Name oxozinc
Molecular Weight (g/mol) 81.38
PubChem CID 14806
ChEBI CHEBI:36560
Formula Weight 81.37
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Zinc oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.