missing translation for 'onlineSavingsMsg'
Learn More

Zinc oxide substrate, 10x10x0.5mm, polished one side, 0001 orientation

Product Code. 15496955
Change view
Click to view available options
Chemical Name or Material:
Zinc Oxide Substrate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Chemical Name or Material Unit Size
15496955 Zinc Oxide Substrate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15496955 Supplier Thermo Scientific Alfa Aesar Supplier No. 045486.KT
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

0001 Orientation | CAS: 1314-13-2 | OZn | 81.38 g/mol

Zinc oxide substrate is one of the suitable substrate for use as wide band gap semiconductor application. It is also used in power device, LED, sensor and detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1314-13-2
Color White
Physical Form Substrate
Quantity 1 Ea.
Molecular Formula OZn
MDL Number MFCD00011300
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
Solubility Information Insoluble in water and alcohol. Soluble in acids and alkalies.
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILES O=[Zn]
IUPAC Name oxozinc
Molecular Weight (g/mol) 81.38
PubChem CID 14806
ChEBI CHEBI:36560
Formula Weight 81.37
Odor Odorless
Chemical Name or Material Zinc Oxide Substrate, Polished One Side
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.