missing translation for 'onlineSavingsMsg'
Learn More

Zinc phthalocyanine, 96%

Product Code. 10595101 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10595101 5 g 5g
10495081 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10595101 Supplier Thermo Scientific Supplier No. 422750050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14320-04-8 | C32H16N8Zn | 577.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14320-04-8
InChI Key PODBBOVVOGJETB-UHFFFAOYSA-N
Synonym zinc phthalocyanine, znpc, phthalocyanine zinc, zinc ii phthalocyanine, phthalocyanine, zinc, zinc phthalocyanine, dye content 97 %
PubChem CID 2735172
ChEBI CHEBI:51218
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Zn+2]

Specifications

Color Purple
Loss on Drying 5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C32H16N8Zn
Quantity 5 g
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 577.91
Formula Weight 577.91
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Zinc phthalocyanine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.