missing translation for 'onlineSavingsMsg'
Learn More

Zinc powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11386557
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
5 x 1 kg
Unit Size:
1kg
250g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386557 5 kg 5kg
11396557 250 g 250g
11306567 1 kg 1kg
15430967 5 x 1 kg 1kg
11316567 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11386557 Supplier Thermo Scientific Alfa Aesar Supplier No. 000424.A7
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Appearance Gray powder
Quantity 5 kg
Mesh Size -100 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Vapor Pressure 1 hPa (1mmHg)
Assay Percent Range 99.9% (Metals basis)
Form Powder
Formula Weight 65.38
Chemical Name or Material Zinc
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.