missing translation for 'onlineSavingsMsg'
Learn More

Zinc powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11396557
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
5 x 1 kg
Unit Size:
1kg
250g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396557 250 g 250g
11306567 1 kg 1kg
15430967 5 x 1 kg 1kg
11386557 5 kg 5kg
11316567 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11396557 Supplier Thermo Scientific Alfa Aesar Supplier No. 000424.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Density 7.14
Quantity 250 g
Mesh Size -100 Mesh
Sensitivity Air and moisture sensitive
Assay Percent Range 99.9% (metals basis)
Form Powder
Formula Weight 65.38
Chemical Name or Material Zinc
compliance-icons compliance-icons
Product Identifier
  • Zinc powder
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
  • Substance which, in contact with water, emits flammable gases Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P422-Store contents under .
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.