missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Residuals , 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Zinc, Residuals , 50g
Specifications
Specifications
| Chemical Name or Material | Tellurium/Antimony/Bismuth/Nickel/Silver/Cobalt/Manganese/Iron/Tin/Magnesium/Cadmium/Copper/Lead/Aluminium |
| CAS | 7440-66-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Tellurium 0.0016 %/Antimony 0.0019 %/Bismuth 0.0043 %/Nickel 0.0047 %/Silver 0.0131 %/Cobalt 0.019 %/Manganese 0.0227 %/Iron 0.0283 %/Tin 0.0547 %/Magnesium 0.0568 %/Cadmium 0.147 %/Copper 0.216 %/Lead 0.492 %/Aluminium 1 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction