missing translation for 'onlineSavingsMsg'
Learn More

Zinc rod, 1.27cm (0.5in) dia, 99.99+% (metals basis)

Product Code. 11365928
Change view
Click to view available options
Quantity:
30 cm
60 cm
90 cm
Unit Size:
30cm
60cm
90cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365928 30 cm 30cm
11385928 60 cm 60cm
11375928 90 cm 90cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365928 Supplier Thermo Scientific Alfa Aesar Supplier No. 010433.BV
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Quantity 30 cm
Odor Odorless
Weight ≈9.03g/cm
Form Rod
Percent Purity ≥99.99%
Assay (metals basis)
Chemical Name or Material Zinc rod, 1.27cm (0.5 in.) dia.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.