missing translation for 'onlineSavingsMsg'
Learn More

Zinc Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical

Product Code. 10264340 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
HDPE plastic bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10264340 100 mL HDPE plastic bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10264340 Supplier Fisher Chemical Supplier No. J/8340/05
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

100mL, HDPE plastic bottle

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Quantity 100 mL
Packaging HDPE plastic bottle
Form Liquid
MDL Number 11291
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.