missing translation for 'onlineSavingsMsg'
Learn More

Zinc Stearate, Technical, Fisher Chemical

Product Code. 10030370 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10030370 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10030370 Supplier Fisher Chemical Supplier No. Z/1400/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 118°C
CAS 557-05-1
Color White
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula C36H70O4Zn
MDL Number 13031
Synonym zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat
InChI Key XOOUIPVCVHRTMJ-UHFFFAOYSA-L
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2]
IUPAC Name zinc;octadecanoate
Molecular Weight (g/mol) 632.332
PubChem CID 11178
Formula Weight 632.34g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.