missing translation for 'onlineSavingsMsg'
Learn More

Zinc stearate, ZnO 12.5-14%

Product Code. 11337548
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337548 50 g 50g
11307548 250 g 250g
11317548 1 kg 1kg
11327548 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11337548 Supplier Thermo Scientific Alfa Aesar Supplier No. 033238.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 557-05-1 | C36H70O4Zn | 632.332 g/mol

Zinc stearate is used as a release agent for the manufacturing of rubber, polyurethane and polyester processing. It is a lubricant and thickening agent in cosmetic industry. It plays a vital role as an activator for rubber vulcanization. It acts as a phase transfer catalyst in the saponification of fats due to its lipophilic character. It is an active component in fanning powder, which is utilized to decrease the friction between cards.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-05-1
Molecular Formula C36H70O4Zn
Molecular Weight (g/mol) 632.332
MDL Number MFCD00013031
InChI Key XOOUIPVCVHRTMJ-UHFFFAOYSA-L
Synonym zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat
PubChem CID 11178
IUPAC Name zinc;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2]

Specifications

Melting Point 130°C
Color White
Quantity 50 g
Merck Index 14,10158
Solubility Information Insoluble in water,alcohol and ether; Soluble in benzeneSoluble in acids,aromatic and chlorinated hydrocarbons; Insoluble in water,alcohol and ether.
Formula Weight 632.48
Physical Form Powder
Chemical Name or Material Zinc stearate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.