missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate, 100mM aq. soln., Thermo Scientific™

Product Code. 15453639
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453639 100 mL 100mL
15443639 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453639 Supplier Thermo Scientific Alfa Aesar Supplier No. J60963.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7733-02-0
Molecular Formula O4SZn
Molecular Weight (g/mol) 161.44
MDL Number MFCD00011302
InChI Key NWONKYPBYAMBJT-UHFFFAOYSA-L
Synonym zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
PubChem CID 24424
ChEBI CHEBI:35176
IUPAC Name zinc;sulfate
SMILES [Zn++].[O-]S([O-])(=O)=O

Specifications

Color Colorless
Physical Form Liquid
Quantity 100 mL
Linear Formula ZnSO4
UN Number UN3082
Formula Weight 161.43
Concentration or Composition (by Analyte or Components) 100mM aq. soln.
Chemical Name or Material Zinc sulfate
compliance-icons compliance-icons
Product Identifier
  • Zinc sulfate, 100mM aqueous solution
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Zinc sulfate heptahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.