missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate, 7-Hydrate, Granular, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15528294 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15528294 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528294 Supplier Avantor J.T.Baker Supplier No. 438301
  • ONLINE EXCLUSIVE PRICE
    211.94 €
    177.05 € / 500g
    Save 34.89 €
    16% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7446-20-0
Physical Form Granules
Quantity 500 g
Molecular Formula H14O11SZn
MDL Number MFCD00149894
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O
IUPAC Name zinc;sulfate;heptahydrate
Molecular Weight (g/mol) 287.54
PubChem CID 62640
ChEBI CHEBI:32312
Formula Weight 287.56
Grade Multi-Compendial/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.97kg
Chemical Name or Material Zinc Sulfate, 7-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.