missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate heptahydrate, 98%

Product Code. 11429733
Change view
Click to view available options
Quantity:
500 g
2500 g
10 kg
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429733 500 g 500g
11439733 2500 g 2500g
11449733 10 kg 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429733 Supplier Thermo Scientific Alfa Aesar Supplier No. A12915.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7446-20-0 | H14O11SZn | 287.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Zinc sulfate heptahydrate is widely used as a coagulant in the production of rayon. It finds an application in the manufacturing of calico printing, wood preservation and lithopone. It is the source of zinc which is supplied in animal feeds, fertilizers and agricultural sprays. It is utilized as electrolytes for zinc plating and surface treating agents. It is also used as a mordant in dyeing, preservative for skin and leather and in medicine as an astringent and emetic.

Solubility
Soluble in water and glycerol.

Notes
Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 100°C
pH 4.4 to 6
Quantity 500 g
Linear Formula ZnSO4·7H2O
UN Number UN3077
Merck Index 14,10159
Solubility Information Soluble in water and glycerol.
IUPAC Name zinc;sulfate;heptahydrate
Formula Weight 287.54 (161.43 Anhydrous)
Percent Purity 98%
Sensitivity Hygroscopic
Density 1.97
Chemical Name or Material Zinc sulfate heptahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc sulfate heptahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Zinc sulfate heptahydrate, Zinc sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.