missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Heptahydrate, ≥99.0%, Reagent Grade, Honeywell™

Product Code. 15648210 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly bottle
Unit Size:
100g
2.5kg
500kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15648210 500 g Poly bottle 500kg
15628210 100 g Poly bottle 100g
15638210 2.5 kg Poly bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15648210 Supplier Honeywell Research Chemicals Supplier No. Z4750500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Melting Point ∼100°C
Color Colorless
pH 4.4 to 5.6 (at 20°C)
Quantity 500 g
Linear Formula ZnSO4 · 7H2O
UN Number UN3077
Formula Weight 287.56g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Poly bottle
Appearance Crystalline
Density ∽1.96g/cu.cm at 20°C
Chemical Name or Material Zinc sulfate heptahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.