missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.0-103.0%, Honeywell Fluka™

Product Code. p-7071826 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Unit Size:
1kg
25kg
5kg
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15671430

Brand: Honeywell-Fluka 144551KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.0-103.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula ZnSO4 · 7H2O
UN Number 3077
Formula Weight 287.56g/mol
Percent Purity 99 to 103.0%
Grade puriss.
Chemical Name or Material Zinc Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.