missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate Monohydrate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Zinc Sulfate Monohydrate |
| CAS | 7446-19-7 |
| Molecular Formula | H2O4S . Zn . H2O |
| Quantity | 1 g |
| Synonym | Zinc Mesosulfate (ZnH2SO5), ZNS 13, Zinc(II) Sulfate Monohydrate |
| InChI Formula | InChI=1S/H2O4S.H2O.Zn/c1-5(2,3)4;;/h(H2,1,2,3,4);1H2; |
| SMILES | O.[Zn].OS(=O)(=O)O |
| IUPAC Name | sulfuric acid;zinc;hydrate |
| Molecular Weight (g/mol) | 179.5 |
| Formula Weight | 179.9071 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction