Learn More
Zinc Sulfate Solution 0.05M (0.05N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Brand: Fisher Chemical J/8490C/90
Code : 23
Additional Details : CAS Number : 7446-20-0 Weight : 0.79336kg Transport : UN number : 3082 Chem class : 9 Pack group : III
Quantity | 6AMP |
---|
Chemical Identifiers
11302 |
Specifications
6AMP | |
Ampoule | |
0°C |
11302 | |
Liquid | |
100°C |