missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate Solution, 0.05M, Solstice

Product Code. 15624180
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15624180 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15624180 Supplier Honeywell-Fluka Supplier No. 344481L
  • ONLINE EXCLUSIVE PRICE
    150.00 €
    135.60 € / 1 L
    Save 14.40 €
    9% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric, 0.05 M ZnSO4

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

CAS 7733-02-0
Quantity 1 L
Molecular Formula O4SZn
Linear Formula ZnSO4
MDL Number MFCD00011302
UN Number NONH for all modes of transport
Synonym zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
InChI Key NWONKYPBYAMBJT-UHFFFAOYSA-L
SMILES [Zn++].[O-]S([O-])(=O)=O
IUPAC Name zinc;sulfate
Molecular Weight (g/mol) 161.44
PubChem CID 24424
ChEBI CHEBI:35176
Formula Weight 161.45g/mol
Chemical Name or Material Zinc sulfate solution
Show More Show Less
compliance-icons
Product Identifier
  • Zinc sulfate solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Zinc sulfate-7-hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.