missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate (Solution in Water), TRC
Specifications
Specifications
| Chemical Name or Material | Zinc Sulfate |
| CAS | 7733-02-0 |
| Quantity | 250 mL |
| Molecular Formula | O4SZn |
| Synonym | Biolectra Zink, Biosanzink, Bonazen, Bufopto Zinc Sulfate, Complexonat, Honny Fresh 10P, Kreatol, Op-Thal-Zin, Optraex, Solvazinc, Solvezink, Sulfuric Acid Zinc Salt, Verazinc, White Vitriol, Z-Span, Zinc Sulfate, Zinc Sulfate (1:1), Zinc Sulfate (ZnSO4), Zinc Vitriol, Zinc(II) Sulfate, Zincaps, Zincate, Zinco, Zincomed, Zincteral |
| InChI Formula | InChI=1S/H2O4S.Zn/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2 |
| SMILES | [Zn+2].[O-]S(=O)(=O)[O-] |
| IUPAC Name | zinc;sulfate |
| Molecular Weight (g/mol) | 161.44 |
| Formula Weight | 159.8809 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?