missing translation for 'onlineSavingsMsg'
Learn More

Zinc Sulfate (Solution in Water), TRC

Product Code. 30456841
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456841 1 L 1 L
30450037 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456841 Supplier Toronto Research Chemicals Supplier No. TRCZ4399851L
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc Sulfate
CAS 7733-02-0
Quantity 1 L
Molecular Formula O4SZn
Synonym Biolectra Zink, Biosanzink, Bonazen, Bufopto Zinc Sulfate, Complexonat, Honny Fresh 10P, Kreatol, Op-Thal-Zin, Optraex, Solvazinc, Solvezink, Sulfuric Acid Zinc Salt, Verazinc, White Vitriol, Z-Span, Zinc Sulfate, Zinc Sulfate (1:1), Zinc Sulfate (ZnSO4), Zinc Vitriol, Zinc(II) Sulfate, Zincaps, Zincate, Zinco, Zincomed, Zincteral
InChI Formula InChI=1S/H2O4S.Zn/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2
SMILES [Zn+2].[O-]S(=O)(=O)[O-]
IUPAC Name zinc;sulfate
Molecular Weight (g/mol) 161.44
Formula Weight 159.8809
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.