missing translation for 'onlineSavingsMsg'
Learn More

Zinc, Technical, Stick (Metal), Low In Arsenic, Fisher Chemical

Product Code. 10558830 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic container
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10558830 500 g Plastic container 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10558830 Supplier Fisher Chemical Supplier No. Z/0550/53
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Gray
CAS 7440-66-6
Melting Point 419°C
Boiling Point 907°C
Quantity 500 g
Packaging Plastic container
Vapor Pressure 1.3mbar 478
Form Solid
Molecular Formula Zn
MDL Number MFCD00011291
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES [Zn]
IUPAC Name zinc
Molecular Weight (g/mol) 65.38
PubChem CID 23994
ChEBI CHEBI:30185
Formula Weight 65.36g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.