missing translation for 'onlineSavingsMsg'
Learn More

Zinc Trifluoroacetate Hydrate, TRC

Product Code. 30450588
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450588 1 g -
30450051 100 mg 100mg
30456911 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450588 Supplier Toronto Research Chemicals Supplier No. TRCZ2179851G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207801-31-8
Molecular Formula Zn(CF3COO)2 . xH2O
Molecular Weight (g/mol) 309.4
IUPAC Name zinc;2,2,2-trifluoroacetate;hydrate
SMILES O.[Zn+2].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/2C2HF3O2.H2O.Zn/c2*3-2(4,5)1(6)7;;/h2*(H,6,7);1H2;/q;;;+2/p-2
Formula Weight 307.9098
Chemical Name or Material Zinc Trifluoroacetate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.