missing translation for 'onlineSavingsMsg'
Learn More

Zinc trifluoromethanesulfonate, 98%

Product Code. 11372817
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11372817

Brand: Thermo Scientific Alfa Aesar L15969.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54010-75-2 | C2F6O6S2Zn | 363.51 g/mol

Zinc trifluoromethanesulfonate acts as a catalyst for the preparation of dithioketals. It is used as a Lewis acid catalyst in silylation reactions. It is also used as a catalyst for greener amine synthesis by reductive amination with hydrogen gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc trifluoromethanesulfonate acts as a catalyst for the preparation of dithioketals. It is used as a Lewis acid catalyst in silylation reactions. It is also used as a catalyst for greener amine synthesis by reductive amination with hydrogen gas.

Solubility
Soluble in water and acetonitrile. Slightly soluble in methanol. Insoluble in dichloromethane.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54010-75-2
Molecular Formula C2F6O6S2Zn
Molecular Weight (g/mol) 363.51
MDL Number MFCD00013229
InChI Key CITILBVTAYEWKR-UHFFFAOYSA-L
Synonym zinc trifluoromethanesulfonate, zinc ii trifluoromethanesulfonate, zinc trifluoromethanesulphonate, zinc triflate, zinc ii triflate, zn otf 2, methanesulfonic acid, trifluoro-, zinc salt, zinc 2+ ion ditriflate, methanesulfonic acid, 1,1,1-trifluoro-, zinc salt 2:1, zinc ii trifluoromethanesulphonate
PubChem CID 104671
SMILES [Zn++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point >300°C
Linear Formula (CF3SO3)2Zn
Quantity 5 g
UN Number UN3261
Beilstein 4028195
Sensitivity Hygroscopic
Solubility Information Soluble in water and acetonitrile. Slightly soluble in methanol. Insoluble in dichloromethane.
IUPAC Name zinc(2+) ditrifluoromethanesulfonate
Formula Weight 363.51
Percent Purity 98%
Chemical Name or Material Zinc trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.