missing translation for 'onlineSavingsMsg'
Learn More

Zinc trifluoromethanesulfonate, 98%

Product Code. 11382817
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382817 25 g 25g
11372817 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382817 Supplier Thermo Scientific Alfa Aesar Supplier No. L15969.14
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 54010-75-2 | C2F6O6S2Zn | 363.51 g/mol

Zinc trifluoromethanesulfonate acts as a catalyst for the preparation of dithioketals. It is used as a Lewis acid catalyst in silylation reactions. It is also used as a catalyst for greener amine synthesis by reductive amination with hydrogen gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc trifluoromethanesulfonate acts as a catalyst for the preparation of dithioketals. It is used as a Lewis acid catalyst in silylation reactions. It is also used as a catalyst for greener amine synthesis by reductive amination with hydrogen gas.

Solubility
Soluble in water and acetonitrile. Slightly soluble in methanol. Insoluble in dichloromethane.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 54010-75-2
Molecular Formula C2F6O6S2Zn
Molecular Weight (g/mol) 363.51
MDL Number MFCD00013229
InChI Key CITILBVTAYEWKR-UHFFFAOYSA-L
Synonym zinc trifluoromethanesulfonate, zinc ii trifluoromethanesulfonate, zinc trifluoromethanesulphonate, zinc triflate, zinc ii triflate, zn otf 2, methanesulfonic acid, trifluoro-, zinc salt, zinc 2+ ion ditriflate, methanesulfonic acid, 1,1,1-trifluoro-, zinc salt 2:1, zinc ii trifluoromethanesulphonate
PubChem CID 104671
SMILES [Zn++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Spécification

Melting Point >300°C
Linear Formula (CF3SO3)2Zn
Quantity 25 g
UN Number UN3261
Beilstein 4028195
Sensitivity Hygroscopic
Solubility Information Soluble in water and acetonitrile. Slightly soluble in methanol. Insoluble in dichloromethane.
IUPAC Name zinc;trifluoromethanesulfonate
Formula Weight 363.51
Percent Purity 98%
Chemical Name or Material Zinc trifluoromethanesulfonate
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Zinc trifluoromethanesulfonate
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Mention de danger
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contain: Zinc trifluoromethanesulfonate

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.