missing translation for 'onlineSavingsMsg'
Learn More

Zinc wire, 0.25mm (0.01in) dia, Puratronic™, 99.994% (metals basis)

Product Code. 11326408
Change view
Click to view available options
Quantity:
2 m
10 m
50 m
Unit Size:
10m
2m
50m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326408 2 m 2m
11346408 10 m 10m
11336408 50 m 50m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326408 Supplier Thermo Scientific Alfa Aesar Supplier No. 012055.G2
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 0.35g/m | CAS: 7440-66-6 | Zn | 65.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Appearance Gray wire
Density 7.14g/cm3
Quantity 2 m
Odor Odorless
Assay Percent Range 99.994% (Metals basis)
Form Wire
Chemical Name or Material Zinc

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.