missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Zn/Al/Cu, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Zinc, Zn/Al/Cu, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Antimony/Tin/Lead/Cadmium/Bismuth/Iron/Manganese/Nickel/Magnesium/Copper/Aluminium |
| CAS | 7440-66-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Antimony 0.0005 %/Tin 0.0017 %/Lead 0.0029 %/Cadmium 0.0036 %/Bismuth 0.0053 %/Iron 0.0053 %/Manganese 0.0067 %/Nickel 0.008 %/Magnesium 0.023 %/Copper 1.17 %/Aluminium 8.3 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction