missing translation for 'onlineSavingsMsg'
Learn More

Zincon monosodium salt

Product Code. 11471307
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471307 25 g 25g
11451307 1 g 1g
11461307 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471307 Supplier Thermo Scientific Alfa Aesar Supplier No. B24256.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 62625-22-3 | C20H16N4NaO6S | 463.42 g/mol

Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zincon monosodium salt is a colorimetric indicator used in the spectrophotometric determination of zinc, mercury and copper. Further, it is used for automated flow injection analysis of zinc.

Solubility
Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-22-3
Molecular Formula C20H16N4NaO6S
Molecular Weight (g/mol) 463.42
MDL Number MFCD00064385
InChI Key IABRINWJUBAIIE-JJECXDOKSA-N
PubChem CID 131856391
IUPAC Name 2-[2-[(Z)-N-[(Z)-(6-oxo-3-sulfocyclohexa-2,4-dien-1-ylidene)amino]-C-phenylcarbonimidoyl]hydrazinyl]benzoic acid;sodium
SMILES C1=CC=C(C=C1)C(=NN=C2C=C(C=CC2=O)S(=O)(=O)O)NNC3=CC=CC=C3C(=O)O.[Na]

Specifications

Melting Point ∼203°C (decomposition)
Quantity 25 g
Beilstein 3872699
Solubility Information Soluble in aqueous alkali. Slightly soluble in water and alcohol. Insoluble in other organic solvents.
Formula Weight 462.42
Chemical Name or Material Zincon monosodium salt
compliance-icons
Product Identifier
  • Zincon monosodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Zincon monosodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.