missing translation for 'onlineSavingsMsg'
Learn More

Zinpyr-1, TRC

Product Code. 30450854
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450854 10 mg -
30455065 5 mg 5mg
30449598 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450854 Supplier Toronto Research Chemicals Supplier No. TRCZ44000010MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288574-78-7
Molecular Formula C46 H36 Cl2 N6 O5
Molecular Weight (g/mol) 823.72
IUPAC Name 2-[4,5-bis[[bis(pyridin-2-ylmethyl)amino]methyl]-2,7-dichloro-3-hydroxy-6-oxoxanthen-9-yl]benzoic acid
SMILES OC(=O)c1ccccc1C2=C3C=C(Cl)C(=O)C(=C3Oc4c(CN(Cc5ccccn5)Cc6ccccn6)c(O)c(Cl)cc24)CN(Cc7ccccn7)Cc8ccccn8

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C46H36Cl2N6O5/c47-39-21-35-41(33-15-1-2-16-34(33)46(57)58)36-22-40(48)43(56)38(28-54(25-31-13-5-9-19-51-31)26-32-14-6-10-20-52-32)45(36)59-44(35)37(42(39)55)27-53(23-29-11-3-7-17-49-29)24-30-12-4-8-18-50-30/h1-22,55H,23-28H2,(H,57,58)
Formula Weight 822.2124
Chemical Name or Material Zinpyr-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.