missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zip Alexa Fluor™ Rapid Antibody Labeling Kits

Product Code. 15982354
Change view
Click to view available options
Label or Dye:
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 647
Excitation/Emission:
490/525 nm
555/580 nm
650/665 nm
Unit Size:
Pack of 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Label or Dye Excitation/Emission Unit Size
15982354 Alexa Fluor 488 490/525 nm Pack of 3
15992354 Alexa Fluor 555 555/580 nm Pack of 3
15902364 Alexa Fluor 647 650/665 nm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 15982354 Supplier Invitrogen™ Supplier No. Z11233
Keep Your Science Moving with 3 for 2
Use promo code 27970
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

Invitrogen Zip Alexa Fluor Rapid Antibody Labeling kits allow you to efficiently and quickly label your precious antibody with Alexa Flour dyes to create an antibody conjugate that is ready to use within 15 minutes with 100% recovery.

Invitrogen Zip Alexa Fluor Rapid Antibody Labeling kits allow you to efficiently and quickly label your precious antibody with Alexa Flour dyes to create an antibody conjugate that is ready to use within 15 minutes with 100% recovery. Each kit contains everything needed for three separate labeling reactions of 100 μg of antibody. Simply add 1 mL water to component A containing Zip buffer, add 100 μg of antibody, incubate for 15 minutes, and your antibody is ready to use. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry (IHC), primary detection, ELISAs, immunocytochemistry (ICC), and indirect FISH.

Create ready-to-use antibody conjugate in just 15 minutes with our Invitrogen Zip Alexa Fluor Rapid Antibody Labeling kits. These kits enable you to efficiently label your precious antibody with various Alexa Flour dyes.

Important features of Zip Alexa Fluor Rapid Antibody Labeling kits:

  • Speed—antibody conjugates ready to use in 15 min
  • Efficiency—100% antibody recovery
  • Versatility—kits are useful for a wide range of applications
  • Easy-to-use—no chemistry or conjugation experience needed
  • Flexibility—choose from Alexa 488, 555, and 647

Each kit contains everything you need to perform three separate labeling reactions of 100 μg of antibody. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry (IHC), primary detection, ELISA, immunocytochemistry (ICC), and indirect FISH.

Benefits of using Zip Alexa Fluor Rapid Antibody Labeling kits:

Specificity and versatility

A primary antibody that is directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Multiple primary antibodies of the same isotype or derived from the same species can be used in the same experiment if they are directly labeled with compatible fluorophores.

Robustness

Compared to traditional dyes, Alexa Fluor dyes are brighter, more photostable, and more pH resistant between pH 4 and 10. When using Alexa Fluor dyes, higher degrees of labeling can usually be achieved without intramolecular quenching.

Customization is possible

Can't find what you're looking for in our stocked list? We can prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeling Kit
Quantity 3 Labelings
Content And Storage Reactive fluorescent label (3 vials), sodium bicarbonate (3 vials). Store desiccated and protected from light in the freezer (-5°C to +30°C).
Excitation/Emission 490/525 nm
Labeling Scale 100 μg
Product Line Alexa Fluor
Chemical Reactivity Amine
Label or Dye Alexa Fluor 488
Color Green
Labeling Target IgG
Label Type Alexa Fluor
Show More Show Less
With the Zip Alexa Fluor Rapid Antibody Labeling kits, can I store the labeled antibody after the 15 mins reaction?

No, it is intended for immediate use. If you wish to store your labeled antibody after the reaction, it should undergo a post-reaction clean-up using either size exclusion resin (such as Bio-Rad BioGel P-30) or Dye Removal Columns (Cat No. 22858).

Why do the Zip Alexa Fluor Rapid Antibody Labeling kits not require a post-reaction purification step?

This is because we have optimized the amount of dye to label 100 µg of antibody per reaction and after the incubation time of 15 mins, most of the reactive dye is either bound or unreactive due to spontaneous hydrolysis, leaving little or no reactive dye to label your sample.

I am planning to use one of the Zip Alexa Fluor Rapid Antibody Labeling kits. My antibody is shipped in Tris buffer with BSA added as a stabilizer. Do I need to remove the BSA or Tris prior to the reaction?

Yes, the BSA and any component with a primary amine (such as the Tris buffer) would react with the amine-reactive Alexa Fluor dyes used in these labeling kits, resulting in less optimal labeling of the antibody. For small volume purification of the antibody, you may use either Nab Protein A/G Spin Kit, 0.2 mL (Cat No. 89950), Protein A/G Magnetic Agarose Beads (Cat No. 78609), or Melon Gel IgG Spin Purification Kit (Cat No. 45206).

For the Zip Alexa Fluor Rapid Antibody Labeling kits, why does the antibody have to be at a 1 mg/mL concentration?

The 1 mg/mL concentration of the antibody (100 µg of antibody in a 100 µL volume) allows for optimal reaction kinetics. A more dilute solution of antibody would exhibit suboptimal labeling due to a greater amount of reactive dye lost to spontaneous hydrolysis; the reactive group on the dyes is labile in aqueous solutions.

What is included in the Zip Alexa Fluor Rapid Antibody Labeling kits?

Each kit contains 3 vials of solid sodium bicarbonate (Zip buffer reagent, Component A), 3 vials of the Alexa Fluor reactive dye (Component B), and a Quick Reference Card. Sufficient reagents are provided to label 100 µg of antibody per reaction (100 µL of a 1 mg/mL antibody solution), 3 reactions per kit.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.