missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zip Alexa Fluor™ Rapid Antibody Labeling Kits

Product Code. 15992354
Click to view available options
Label or Dye:
Alexa Fluor 488
Alexa Fluor 555
Alexa Fluor 647
Excitation/Emission:
490/525 nm
555/580 nm
650/665 nm
Unit Size:
Pack of 3
This item is not returnable. View return policy

Product Code. 15992354

Brand: Invitrogen™ Z11234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Invitrogen Zip Alexa Fluor Rapid Antibody Labeling kits allow you to efficiently and quickly label your precious antibody with Alexa Flour dyes to create an antibody conjugate that is ready to use within 15 minutes with 100% recovery.

Invitrogen Zip Alexa Fluor Rapid Antibody Labeling kits allow you to efficiently and quickly label your precious antibody with Alexa Flour dyes to create an antibody conjugate that is ready to use within 15 minutes with 100% recovery. Each kit contains everything needed for three separate labeling reactions of 100 μg of antibody. Simply add 1 mL water to component A containing Zip buffer, add 100 μg of antibody, incubate for 15 minutes, and your antibody is ready to use. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry (IHC), primary detection, ELISAs, immunocytochemistry (ICC), and indirect FISH.

Create ready-to-use antibody conjugate in just 15 minutes with our Invitrogen Zip Alexa Fluor Rapid Antibody Labeling kits. These kits enable you to efficiently label your precious antibody with various Alexa Flour dyes.

Important features of Zip Alexa Fluor Rapid Antibody Labeling kits:

  • Speed—antibody conjugates ready to use in 15 min
  • Efficiency—100% antibody recovery
  • Versatility—kits are useful for a wide range of applications
  • Easy-to-use—no chemistry or conjugation experience needed
  • Flexibility—choose from Alexa 488, 555, and 647

Each kit contains everything you need to perform three separate labeling reactions of 100 μg of antibody. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry (IHC), primary detection, ELISA, immunocytochemistry (ICC), and indirect FISH.

Benefits of using Zip Alexa Fluor Rapid Antibody Labeling kits:

Specificity and versatility

A primary antibody that is directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Multiple primary antibodies of the same isotype or derived from the same species can be used in the same experiment if they are directly labeled with compatible fluorophores.

Robustness

Compared to traditional dyes, Alexa Fluor dyes are brighter, more photostable, and more pH resistant between pH 4 and 10. When using Alexa Fluor dyes, higher degrees of labeling can usually be achieved without intramolecular quenching.

Customization is possible

Can't find what you're looking for in our stocked list? We can prepare a custom antibody conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Kit Contents Reactive fluorescent label (3 vials), sodium bicarbonate (3 vials)
Product Type Labeling Kit
Quantity 3 Labelings
Content And Storage Store desiccated and protected from light in the freezer (-5°C to +30°C).
Excitation/Emission 555/580 nm
Labeling Scale 100 μg
Product Line Alexa Fluor
Chemical Reactivity Amine
Label or Dye Alexa Fluor 555
Color Orange
Labeling Target IgG
Label Type Alexa Fluor
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.