Learn More
Chemsplash Zip Labcoat T5/6 Fabric, Elastic Cuff
GREENER_CHOICE
Product Code. 30058202
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30058202 Small Pack of 50
30058450 Medium Pack of 50
30058447 Large Pack of 50
30058301 X-Large Pack of 50
30058165 2X-Large Pack of 50
30058354 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30058202 Supplier Chemsplash Supplier No. 2549.400.004
  • ONLINE EXCLUSIVE PRICE
    231.00 €
    228.18 € / Pack of 50
    Save 2.82 €
    1% Off
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Lab Coat is a 2/3 length coat with a collar and zip closure and elasticated wrists. It is anti-static to meet with EN1149-5:2008, lint free and suitable for clean room applications.
Chemsplash Lab Coat
  • Chemsplash 55GSM Type PB 6B Fabric
  • Zip Fasten Front with Adhesive Flap Cover
  • Elasticated Cuffs
  • Fold Over Collar
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • PFAS Free
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
Description: The Chemsplash Lab Coat is made from Chemsplash fabric, which is soft and microporous. It is strong and resistant to chemical splash and dust. This lab coat is a 2/3 length coat with a collar, zip closure, and elasticated wrists. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from the environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifications
Product Type Lab Coat
Size Small
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Liquids
No. of Pockets None
Latex Free Yes
Autoclavable Autoclavable
Cleanroom Class Class 100-100,000 (ISO 5-8)