Learn More
Chemsplash Zip Labcoat T5/6 Fabric, Elastic Cuff
Product Code. 30058426
Change view
Click to view available options
Size:
4X-Large
5X-Large
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058426 4X-Large Pack of 50
30058255 5X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30058426 Supplier Chemsplash Supplier No. 2771.400.010
  • ONLINE EXCLUSIVE PRICE
    268.38 €
    249.09 € / Pack of 50
    Save 19.29 €
    7% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Chemsplash Lab Coat is made from Chemsplash fabric which is soft and microporous. This lab coat is a 2/3 length coat with a collar and zip closure and elasticated wrists.
Chemsplash Lab Coat
  • 55GSM Type PB 6B Fabric
  • Zip Fasten Front with Adhesive Flap Cover
  • Elasticated Cuffs
  • Fold Over Collar
  • Anti-Static Material
  • Non-Linting
  • Silicone and Latex Free
  • PFAS Free
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Lab Coat is made from Chemsplash fabric, which is soft, microporous, and resistant to chemical splash and dust. This lab coat features a 2/3 length design with a collar and zip closure, along with elasticated wrists for a snug fit. It is anti-static to meet EN1149-5:2008 standards, lint-free, and suitable for cleanroom applications. The lab coat conforms to ISO 14644-1:2015 Cleanroom Class 5-9, ensuring it provides the protection required for sensitive environments. Chemsplash accessories are designed to protect both the wearer and the environment, offering maximum comfort and wearability.
Specifications
Product Type Lab Coat
Size 4X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Liquids
No. of Pockets None
Latex Free Yes
Autoclavable Autoclavable
Cleanroom Class Class 100-100,000 (ISO 5-8)