missing translation for 'onlineSavingsMsg'
Learn More

Merck ZipTip™ Pipet Tips

Sample preparation and precise liquid handling in one device

Brand:  Merck ZTC18S960

Code : 72

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 10044590

  • 4370.00 € / Pack of 960
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

Sample preparation and precise liquid handling in one device

A 0.6μL bed of C18, C4, or SCX (strong cation exchange) media is fixed at the end of a 10μL pipet tip such that there is no dead volume; concentrated, purified samples are eluted in 1 to 4μL. For applications requiring smaller elution volumes (e.g., <1μL), ZipTip μ-C18 tips feature a microbed format containing 0.2μL of media.

  • Concentrate and purify 1–100μL biological samples
  • Ideal for purifying and concentrating femtomoles to picomoles of peptides, proteins, or oligonucleotides in minutes prior to mass spectrometry, HPLC, capillary electrophoresis, and other analytical techniques

Specifications

Specifications

With C18 Media
1.25 in.
Specialty, Media-Fixed
Universal Fit
Polypropylene
10 μL
3.1cm
960/Pk.
ZipTip
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers