missing translation for 'onlineSavingsMsg'
Learn More

Zirconium foil, 0.25mm (0.01in) thick, annealed, 99.8% (metals basis excluding Hf), Hf nominal 2%, Thermo Scientific Chemicals

Product Code. 15464735
Change view
Click to view available options
Quantity:
100 x 125 mm
125 x 500 mm
125 x 2000 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464735 125 x 2000 mm Each
11375799 100 x 125 mm Each
11300759 125 x 500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15464735 Supplier Thermo Scientific Alfa Aesar Supplier No. 010591.IB
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Form: Approx.20.6g/100 x 125mm, Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Color Gray to Silver
Appearance Gray to silver foil
Quantity 125 x 2000 mm
Odor Odorless
Assay Percent Range 99.8% (Metals basis excluding Hf)
Form Foil
Chemical Name or Material Zirconium, Annealed

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.