Learn More
Zirconium(IV) 2,4-pentanedionate, Thermo Scientific Chemicals
Descripción
Zirconium(IV) 2,4-pentanedionate is used as a cross-linking agent for oxygen-containing polymers. It is used as a catalyst in organic synthesis for the nitration of o-nitrotoluene using a dinitrogen pentoxide to get 2,4-dinitrotoluenes and 2,6-dinitrotoluenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Melting Point | 191°C to 195°C |
| Color | White |
| Odor | Weak |
| Quantity | 50 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 487.66 |
| Physical Form | Crystals |
| Chemical Name or Material | Zirconium(IV) 2,4-pentanedionate |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.