missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) bromide, 99%, Thermo Scientific Chemicals

Product Code. 11324949
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324949 10 g 10g
11334949 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324949 Supplier Thermo Scientific Alfa Aesar Supplier No. 086139.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Zirconium(IV) bromide is used as a starting material in the synthesis of zirconium hydride complexes as well as unique cluster compounds. It is also used as a biochemical for proteomics research and as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13777-25-8
Molecular Formula Br4Zr
Molecular Weight (g/mol) 410.84
MDL Number MFCD00049637
InChI Key LSWWNKUULMMMIL-UHFFFAOYSA-J
Synonym zirconium tetrabromide, zrbr4, zirconium iv bromide, zirconium bromide zrbr4 , t-4, zirconium bromide zrbr4, zirconium bromide zrbr4 8ci 9ci, zirconium iv bromide, anhydrous trace metals basis
PubChem CID 83727
IUPAC Name tetrabromozirconium
SMILES [Br-].[Br-].[Br-].[Br-].[Zr+4]

Specifications

Melting Point 450° (15 atm)
Physical Form Powder
Quantity 10 g
Assay Percent Range 99%
Solubility Information Soluble in water.
Formula Weight 410.86
Sensitivity Moisture sensitive
Density 4.201
Chemical Name or Material Zirconium(IV) Bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.